Learn More
MSC Sealed Air Instapak Quick Bag, 18 x 24 in, 30/Case
Catalog No. 501918373
Encompass
Instapak Quick® RT expandable foam bags provide instant, protective foam cushions at room temperature. This foam-in-a-bag system activates chemicals directly inside the bag, eliminating the need for preheating or cleanup.
  • Easy to use, protective foam cushions
  • Completely portable, no preheating warming unit required
  • Recommended temperature range is 65° - 90° F (18.3° - 32.2° C)
  • Bag #40, suitable for 12 × 12 × 12 in boxes
Catalog No. 50-191-8373 Supplier MSC Supplier No. 39579602
Instapak Quick® RT expandable foam bags provide instant, protective foam cushions at room temperature. This foam-in-a-bag system activates chemicals directly inside the bag, eliminating the need for preheating or cleanup.
  • Easy to use, protective foam cushions
  • Completely portable, no preheating warming unit required
  • Recommended temperature range is 65° - 90° F (18.3° - 32.2° C)
  • Bag #40, suitable for 12 × 12 × 12 in boxes