Learn More
BINDER™ Calibration Certificate, Expanded
SDP
Catalog No. 08112021 Shop All BINDER Inc. Products
Change view
Click to view available options
For Use With (Equipment):
BINDER Chambers Series MK
Catalog No. For Use With (Equipment)
08-112-021 BINDER Chambers Series MK
1 options
Catalog No. 08-112-021 Supplier BINDER™ Supplier No. 80121124
Add to Cart
Add to Cart
Calibration certificate, expanded - for temperature; for extending the measurement in center of chamber to include another test temperature
  • Each additional measurement at an additional measuring point or set of values
  • Extension
Specifications
For Use With (Equipment) BINDER Chambers Series MK
Description Calibration certificate, expanded - for temperature; for extending the measurement in center of chamber to include another test temperature
Calibration Points 1 Measuring Point
Duration 60 to 120 min.
Calibration Type Temperature Measurement
Product Type Calibration Certificate (Expanded)