Learn More
Sigma Aldrich Fine Chemicals Biosciences Expanded tabletop plasma c1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z5616571EA
Encompass
A larger tabletop plasma cleaner with four times the capacity of the basic plasma cleaner suitable for nanoscale surface cleaning and activation of small samples. Plasma treatment may be applied to a wide variety of materials including glass semiconductors polymers and metals. Useful applications include surface cleaning of substrates for enhanced adhesion plasma activation for microfluidic device fabrication and surface chemistry modification of biomaterials and polymers..nbspAdjustable RF power (Low Medium High power settings) Maximum 30W.nbsp6 in. diameter x 6.5 in length Pyrex chamber (optional quartz chamber).nbspHinged door with viewing window.nbspActive fan cooling.nbsp1/8 NPT metering valve to qualitatively control gas flow and chamber pressure.nbsp1/8 NPT 3-way valve to quickly switch from introducing gas venting and isolating the chamber.nbspRequires a separate vacuum pump to run the plasma process
Catalog No. 50-290-8567
Add to Cart
Request a Quote