Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Expanding Science Skills Series - Chemistry Book
SDPSCI_ED

Catalog No. S02431A
Change view
Click to view available options
Grade:
6 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S02431A 6 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02431A Supplier Carson Dellosa Publishing Supplier No. CD404119
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specializes in providing captivating, supplemental books and decorative resources to complement middle- and upper-grade classrooms.

Product line covers mathematics, sciences, language arts, social studies, history, government, fine arts, and character.

  • Designed by leading educators
  • Supports NSE, NCTM Standards

Specifications

Grade 6 to 12
Format Book
Product Type Chemistry Resource Book
For Use With (Application) Specializes in providing captivating, supplemental books and decorative resources to complement middle and upper grade classroom
Subject Chemistry
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.