Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Expanding Science Skills Series - Earth Science Book
SDPSCI_ED

Catalog No. S02431G
Change view
Click to view available options
Grade:
6 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S02431G 6 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02431G Supplier Carson Dellosa Publishing Supplier No. CD404125
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Product line covers mathematics, sciences, language arts, social studies, history, government, fine arts, and character.

  • Designed by leading educators
  • Supports NSE, NCTM and Standards for Technological Literacy

Specifications

Grade 6 to 12
Format Print and e-Book
Product Type Astronomy Resource Book
Includes Subject specific concepts and terminology, inquiry-based activities, challenge questions, extension activities, assessments, curriculum resources, a bibliography and materials lists
For Use With (Application) Connect students in grades 5 and up with science using Astronomy, It allows for differentiated instruction and supports national science education standards and NCTM standards
Subject Science
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.