Learn More
Sigma Aldrich Fine Chemicals Biosciences ExpandTM High fidelity PCR
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 11759078001
Encompass
The 5 - 3 polymerase activity of Taq DNA Polymerase and the 3 - 5 exonuclease of the proofreading polymerase combine for improved performance and results 3 - 5 exonuclease proofreading produces a threefold increase in DNA synthesis fidelity compared to Taq DNA polymerase.
Catalog No. 50-291-4633