Learn More
Julabo USA Inc Each, 1L expansion vessel for Forte HT units
SDP
Catalog No. 507534176
Encompass_Preferred
Bath Accessories, Julabo, Expansion vessel for Forte HT high temperature circulator models HT30/HT60, connection M16x1. Filling volume 1L. Connection M16x1 male.
Catalog No. 50-753-4176 Supplier Julabo USA Inc Supplier No. 8970801
Bath Accessories, Julabo, Expansion vessel for Forte HT high temperature circulator models HT30/HT60, connection M16x1. Filling volume 1L. Connection M16x1 male.