Learn More
Research Products International Corp Expansion Kit for Roto-Shake Genie, Six Plates with 72 Clips for 10-13 mm Tubes
SDP
Catalog No. 501977629
Encompass
Pack of 6 metal clip plates to accommodate up to 12 tubes with a diameter of 10-13mm.
A maximum of 8 clip plates, 4 per side, may be attached to the Magnetic Platform for use with the Roto-Shake Genie, Enviro-Genie, or Incubator Genie. A maximum of 12 clip plates, 6 per side, may be attached to the Magnetic Platform for use with the Rotator Genie.
Catalog No. 50-197-7629 Supplier Research Products International Corp Supplier No. 248817
Pack of 6 metal clip plates to accommodate up to 12 tubes with a diameter of 10-13mm.
A maximum of 8 clip plates, 4 per side, may be attached to the Magnetic Platform for use with the Roto-Shake Genie, Enviro-Genie, or Incubator Genie. A maximum of 12 clip plates, 6 per side, may be attached to the Magnetic Platform for use with the Rotator Genie.