Learn More
Research Products International Corp Expansion Kit for Roto-Shake, 6 Plates w/ 36 Clips
SDP
Catalog No. 501977627
Encompass
Pack of 6 metal clip plates to accommodate up to 6 tubes with a diameter of 15-17mm.
A maximum of 8 clip plates, 4 per side, may be attached to the Magnetic Platform for use with the Roto-Shake Genie, Enviro-Genie or Incubator-Genie. A maximum of 12 clip plates, 6 per side, may be attached to the Magnetic Platform for use with the Rotator Genie.
Catalog No. 50-197-7627 Supplier Research Products International Corp Supplier No. 248818
Pack of 6 metal clip plates to accommodate up to 6 tubes with a diameter of 15-17mm.
A maximum of 8 clip plates, 4 per side, may be attached to the Magnetic Platform for use with the Roto-Shake Genie, Enviro-Genie or Incubator-Genie. A maximum of 12 clip plates, 6 per side, may be attached to the Magnetic Platform for use with the Rotator Genie.