Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gilson™ EXPERTTM Standard TOWERPACKTM Reload System Pipette Tips
GREENER_CHOICE

Catalog No. F1735051G Shop All Gilson Products
Change view
Click to view available options
Graduations:
10, 50, 100μL
2, 5, 10μL
200, 500, 1000μL
Length (Metric):
4.534cm
5.144cm
10.22cm
Volume (Metric):
0.1 to 10 μL
1 to 200 μL
50 to 1250 μL
Model:
E1000 XL
E200
EL10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Graduations Length (Metric) Volume (Metric) Model
F1735051G 200, 500, 1000μL 10.22cm 50 to 1250 μL E1000 XL
F1732051G 2, 5, 10μL 4.534cm 0.1 to 10 μL EL10
F1733051G 10, 50, 100μL 5.144cm 1 to 200 μL E200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. F1735051G Supplier Gilson™ Supplier No. F1735051
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Feature patented, low-retention polymer that creates hydrophobic surface to minimize sample binding while dispensing

  • 10 layers of tips snap with ease into Tipack boxes
  • Clear packaging allows easy identification of tips
  • Color-coded tip packaging for volume color matching to pipettes
  • Graduations tips enable quick-checks when aspirating samples
  • Economical and easy-to-use
  • Manufactured in sterile, clean-room environment
  • Free of Human DNA, RNase, DNase, PCR Inhibitors and Pyrogens

Specifications

Description Standard TowerPack Reload System Pipette Tips
Volume (Metric) 50 to 1250 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Autoclavable Autoclavable
Model E1000 XL
Length (Metric) 10.22cm
Clean Claims RNase free, DNase free, Human DNA free, Endotoxins free, PCR Inhibitors free
For Use With (Application) Pipetting
Format Rack
Tip Style Standard
Packaging Racked
Quantity 768/Cs.
Product Type Air Displacement Tips
Color Code Blue
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated Yes
Product Line EXPERT
Material Polypropylene
Shape Conical
Graduations 200, 500, 1000μL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.