Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Expi293™ Expression Medium

Catalog No. p-7075864
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 L
10 L
20 L
6 x 1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1435103 10 L
A1435104 20 L
A1435101 1 L
A1435102 6 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. A1435103 Supplier Gibco™ Supplier No. A1435103
Only null left
Add to Cart
Edge
Add to Cart

Expi293 Expression Medium designed for growth and transfection of suspension-adapted human embryonic kidney (HEK) 293 cells.

Expi293 Expression Medium is a chemically defined, serum-free, protein-free medium for growth and transfection of suspension-adapted human embryonic kidney (HEK) 293 cells. It is specifically designed as a core component to support high density culture of Expi293F cells in the Expi293 Expression System for scalable transient protein expression.

  • Supports growth of suspension 293 cultures to densities of over 1.5 x 10^7 cells/mL
  • Transfection compatible serum-free suspension culture medium enables transfection efficiencies of greater than 70% using ExpiFectamine transfection reagent
  • Enables sustained, high level expression of high density transiently transfected cultures, achieving yields of up to 1 gram per liter of recombinant protein
  • Supports growth and transfection of 293 cells in culture formats of less than 1 mL in multiwell plates to greater than 10 L in disposable bioreactors
  • Does not contain phenol red

Expi293 Expression Medium is formulated with GlutaMAX-I reagent. It is ready to use, with no supplementation required. The chemically defined formulation results in high reproducibility and lot-to-lot reliability. Expi293 Expression Medium contains no human or animal-origin components. The medium is not recommended for adherent cell cultures.

TRUSTED_SUSTAINABILITY

Specifications

Cell Line Expi293F™
Classification Animal Origin-free, Chemically-defined, Protein-free, Serum-free
Form Liquid
Product Type Expression Medium
With Additives GlutaMAX
Without Additives No Phenol Red
Product Line Expi293
Quantity 10 L
Content And Storage One 10L Universal Bag
Store in the dark at 2-8°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.