Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Expi293™ Expression Medium
GSA_VAGREENER_CHOICE

Catalog No. a1435102
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 L
10 L
20 L
6 x 1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1435102 6 x 1 L
A1435103 10 L
A1435104 20 L
A1435101 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. A1435102 Supplier Gibco™ Supplier No. A1435102
Only null left
Add to Cart
Edge
Add to Cart

Expi293 Expression Medium designed for growth and transfection of suspension-adapted human embryonic kidney (HEK) 293 cells.

Expi293 Expression Medium is a chemically defined, serum-free, protein-free medium for growth and transfection of suspension-adapted human embryonic kidney (HEK) 293 cells. It is specifically designed as a core component to support high density culture of Expi293F cells in the Expi293 Expression System for scalable transient protein expression.

  • Supports growth of suspension 293 cultures to densities of over 1.5 x 10^7 cells/mL
  • Transfection compatible serum-free suspension culture medium enables transfection efficiencies of greater than 70% using ExpiFectamine transfection reagent
  • Enables sustained, high level expression of high density transiently transfected cultures, achieving yields of up to 1 gram per liter of recombinant protein
  • Supports growth and transfection of 293 cells in culture formats of less than 1 mL in multiwell plates to greater than 10 L in disposable bioreactors
  • Does not contain phenol red

Expi293 Expression Medium is formulated with GlutaMAX-I reagent. It is ready to use, with no supplementation required. The chemically defined formulation results in high reproducibility and lot-to-lot reliability. Expi293 Expression Medium contains no human or animal-origin components. The medium is not recommended for adherent cell cultures.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Cell Line 293E (HEK), 293 (HEK), Expi293F™, FreeStyle™ 293-F Cells, 293F
Cell Type Embryonic Kidney Cells
Classification Animal Origin-free, Chemically-defined, Protein-free, Serum-free
Concentration 1 X
Form Liquid
Product Type Expression Medium
Serum Level Serum-free
Sterility Sterile
With Additives GlutaMAX, Sodium Pyruvate, Sodium Bicarbonate, Dextrose, Pyridoxine HCl
Without Additives No HEPES, No Phenol Red, No Hypoxanthine, No Thymidine, No Insulin, No Vitamin A, No 2-Mercaptoethanol
Product Line Expi293
Quantity 6 x 1 L
Endotoxin Level < 1 EU/mL
Format Bottle(s)
Included Antibiotics No antibiotics
Inorganic Salts Calcium, Magnesium, Sodium Phosphate
Osmolality 275-295 mOsm/kg
Purity or Quality Grade Cell Culture Grade
Content And Storage Store in the dark at +2°C to +8°C
Culture Environment CO2
Culture Type Suspension Cell Culture
Show More Show Less
Can I purify my protein from Expi293 Expression Medium using ProBond or Ni-NTA purification systems?

Expi293 Expression Medium is not directly compatible with ProBond or Ni-NTA purification systems. We recommend performing a buffer exchange or dialyzing the samples before His-tag purification.

Can I use a different 293 cell line with the Expi293 Expression System?

Other 293 cell lines may be used with the Expi293 Expression System. However, before these cell lines may be used for transfection studies, they must be adapted to serum-free, suspension culture in Expi293 Expression Medium and evaluated for transfection and expression.

Which vector do you recommend using with the Expi293 Expression System?

We recommend using the pcDNA 3.4-TOPO TA vector (Cat. No. A14697). This vector contains the native, full-length CMV promoter and a WPRE (Woodchuck Posttranscriptional Regulatory Element) downstream of the cloning site, both of which contribute to high level gene expression (about 2-3 fold higher expression than with pcDNA3.3 TOPO vector, which in turn provides 2-5 fold higher expression than with a standard pcDNA vector). Of course, the expression level is also protein-dependent.

Is the Expi293 Expression System completely animal-origin free (AOF)?

All the components of the system are animal-origin free except for the Opti-MEM I Reduced Serum Medium that is serum-free but not animal-origin free. Please see the Application Note for using the Expi293 Expression System under animal origin-free conditions:
http://www.thermofisher.com/content/dam/LifeTech/migration/files/proteins-expression-isolation-analysis/pdfs.par.5943.file.dat/expi-293-animal-origin-free-co25751.pdf

I am interested in a mammalian expression system that will allow me to obtain very high yields of my protein. What kind of systems do you offer?

For stable high-yield expression, we offer the Freedom CHO-S and Freedom DG44 kits. For transient high-yield expression, we offer the ExpiCHO Expression System, Expi293 Expression System, FreeStyle 293 Expression System, FreeStyle Max 293 Expression System, and FreeStyle Max CHO Expression System. For high-yield expression of functional membrane proteins in Exp293F cells, we offer the Expi293 MembranePro Expression System (Cat. Nos. A25869, A25870) that combines the scalability and ease of use of Expi293 and the technology of MembranePro to allow an increase of more than 20-fold in membrane protein yield compared to the standard, adherent culture MembranePro Functional Protein Expression System. Please see the Application Note (http://www.thermofisher.com/content/dam/LifeTech/global/life-sciences/ProteinExpressionAnalysis/pdfs/PG1391-PJ6257-CO28704-Expi293Adaption-MembraneProTech_AppNoteGlobal-FHR.pdf) for more details.

Do I need to change media after transfection of Expi293 cells?

No. The Expi293 Expression System is designed to run without media exchanges. There is no need to remove transfection complexes or to change growth medium following transfection.

With the Expi293 Expression System, for how many days should I wait before harvesting my protein?

The optimal expression time will be different for each protein, but most tested proteins fall within a 3-7 day window. Since the system scales well, it is recommended that you run a small-scale pilot experiment to determine when to harvest your protein of interest prior to scaling up.

If I let my Expi293 cells grow to a higher density than recommended, will they be okay?

While Expi293 medium can support much higher cell densities, we do not recommend growing your Expi293 cultures beyond 5-6 x 10e6 cells/mL as subsequent transfection and protein expression efficiencies may be reduced. At higher densities, there is also the increased possibility of reaching the point of reduced culture viability. If your seed culture does exceed 5-6 x 10e6 cells/mL, passage the cells once or twice as detailed in the manual, monitoring them for viability and growth rate. Perform a test transfection using the Protein Expression control IgG or your own expression construct of known yield to determine if cell expression performance has been impacted.

My Expi293 Expression Medium (Cat. No. A1435102) was accidentally frozen. Can I still use it?

This product should be stored at 2-8 degrees C and should not be frozen. The biggest issue with accidentally freezing this product is its solubility. If this product was accidentally frozen, we recommend placing it in a 2-8 degree environment and allowing it to slowly thaw overnight. If the product is fully thawed and shows no signs of precipitation, then it should still be usable, but we cannot guarantee effectiveness.

Does Expi293 Expression Medium contain antibiotics?

Expi293 Expression Medium does not contain antibiotics. We do not recommend the use of antibiotics, however, 5 mL/L of Antibiotic-Antimycotic (Cat. No. 15240) containing penicillin, streptomycin, and amphotericin B may be used when required, as stated on page 1 in the manual (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/expi293_exp_medium_man.pdf).

Do I need to remove Anti-Clumping Agent and Pluronic F-68 from the medium while growing Expi293 cells? How about during transfection of these cells?

You can have Anti-Clumping Agent and Pluronic F-68 in the medium while growing Expi293 cells. You need to remove Anti-Clumping Agent at least 2 passages prior to transfection of Expi293 cells because it interferes with transfection. You can add it back to the medium after transfection. Pluoronic F-68 can be present during transfection as it does not interfere with transfection.

Do I need to change media after transfection in the Expi293 Expression System?

No. The Expi293 Expression System is designed to run without media changes. There is no need to remove transfection complexes or to change growth medium following transfection.

I only added one enhancer and not the other with the Expi293 system. Will I still get good protein expression?

The Enhancers are designed to work together for maximal expression. Addition of just one Enhancer will result in reduced expression and may be anywhere from one third to two thirds the level of expression obtained if both Enhancers had been added on time.

What method do you recommend for quantifying the rabbit Antibody-Expressing Positive Control Vector (Cat. No. A14662)?

The formation of intact IgG molecules may be quantitated using a sandwich ELISA designed to capture and detect rabbit IgG. Besides the rabbit IgG positive control, reagents, and consumables that are included in the kit, you will also need purified rabbit IgG to be used as a standard, F(ab')2 goat anti-rabbit IgG HRP conjugate (Cat. No. A10547), Protein A-coated plates (Cat. No. 15130 for clear plates used in colorimetric detection), TMB colorimetric substrate (Cat. No. 34021), SuperBlock (TBS) Blocking Buffer (Cat. No. 37581), and PBS or TBS buffer for washes. There is an example procedure in our Protein A-coated plates manual (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0011310_Pierce_ProteinA_G_AG_Coat_96Well_UG.pdf). Please note, our R&D scientists determine titer values from crude cell culture supernatants using a Pall Life Sciences FortéBio Octet instrument equipped with a protein A biosensor.


WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.