Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Expi293™ Met (–) Protein Labeling Kit

Catalog No. A41249
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A41249 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A41249 Supplier Gibco™ Supplier No. A41249
Get 15% Off Cell Culture Products
Use promo code CELLS15
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Expi293 Met (-) Protein Labeling Kit includes Expi293 Met (-) Expression Medium and the ExpiFectamine 293 Met (-) Transfection Kit for rapid, high-yield transient protein expression from Expi293F cells for applications where labeling of methionine residues in proteins is desired

Kit components are specifically formulated without methionine, allowing users to add labeled methionine of their choice

Features:

  • Methionine-free medium and transfection enhancer formulations
  • 2 to 10-fold higher protein yields compared to other transfection reagents used for high-density 293 cell culture
  • Same transient transfection protocol as ExpiFectamine 293 Transfection Kit
  • Scalable transfection from small to large scale while maintaining equivalent volumetric protein yields

Specifically designed to easily integrate with other components of the Expi293 Expression System, including Expi293F cells and Expi293 Expression Medium

Components included:

  • 1 L Expi293 Met (–) Expression Medium
  • 1 ExpiFectamine 293 Met (–) Transfection Kit sufficient to transfect 1 L of culture
TRUSTED_SUSTAINABILITY

Specifications

Product Type Protein Labeling Kit
Quantity 1 kit
Sufficient For Protein Production
Content And Storage
  • 3 x 0.9 mL ExpiFectamine 293 Transfection Reagent, store at 2°C to 8°CC
  • 5 mL ExpiFectamine 293 Enhancer 1, store at 2°C to 8°CC
  • 50 mL ExpiFectamine 293 Met (–) Enhancer 2, store at 2°C to 8°CC
  • 100 mL Opti-Plex Complexation Buffer, store at 2°C to 8°CC (protect from light)
  • 1 L Expi293 Met (–) Expression Medium, store at 2°C to 8°CC (protect from light)

Shipping Condition Approved for shipment at Room Temperature or on Wet or Dry Ice
Product Line Expi293F
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.