Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Expi293™ PRO Transfection Kit

Catalog No. A40002927
Encompass_Preferred
Change view
Click to view available options
Sufficient For:
For 1 L transfection
For 10 L transfection
For 50 L transfection
For 6 L transfection
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sufficient For
A40002927 For 10 L transfection
A40002926 For 1 L transfection
A40002929 For 6 L transfection
A40002930 For 50 L transfection
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. A40002927 Supplier Gibco™ Supplier No. A40002927
Only null left
Add to Cart
Edge
Add to Cart
Request a Quote

The Expi293 PRO Transfection Kit enables high-efficiency delivery of DNA into HEK293 cells, supporting fast, scalable protein expression. Optimized for use with Expi293F cells, it delivers robust performance across a broad range of protein types.

The Expi293 PRO Transfection Kit is a core component of the Expi293 PRO Expression System. It is designed for transient transfection of high density cultures of HEK 293 cells. The high-efficiency, cationic, lipid-based transfection reagent and transfection enhancers are designed to power the highest possible level of protein expression from Expi293 PRO cells cultured in Expi293 PRO Expression Medium.

  • Designed specifically for transfection of high-density suspension cell culture, with complimentary feed and enhancers that boost transfection performance and protein expression
  • Achieves protein yields 2- to 10-fold higher than other transfection reagents used on high-density Expi293 PRO cell cultures
  • Provides robust and reproducible transfection results, giving users greater confidence in their results
  • Scale transfections for culture volumes of less than 1 mL to greater than 10 liters, while maintaining equivalent volumetric protein yields

The Expi293 PRO Transfection Kit is specifically designed to work with Expi293 PRO cells in the Expi29 PRO Expression System. The transfection kit consists of the cationic lipid-based Expi293 PRO transfection reagent and Expi293 PRO enhancer and feed, which boost transfection, cell viability, and protein expression.

TRUSTED_SUSTAINABILITY

Specifications

Cell Type HEK293
Format Kit
Sample Type Plasmid DNA
Promoter pcDNA™ 3.4
Transfection Technique Lipid-based Transfection, Transient Transfection
Product Line Expi293 PRO
Product Type Transfection Kit
Purity Cell Culture Grade
Quantity For 10 L transfection
Sufficient For For 10 L transfection

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.