Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Expi293F™ GnTI- Cells

Catalog No. A39240
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A39240 1 vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A39240 Supplier Gibco™ Supplier No. A39240
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Expi293F GnTI- cell line is derived from engineered Expi293F cells that do not have N-acetylglucosaminyltransferase I (GnTI) activity and therefore lack complex N-glycans

Powerful tool for the high-yield expression of homogeneously glycosylated recombinant proteins.

Features:

  • Suspension, high-density culture in Expi293 Expression Medium
  • Homogeneous N-glycosylation of expressed proteins
  • Equivalent protein yields to parental Expi293F cells—up to 1 g/L
  • Significantly higher protein yields than other HEK293 GnTI- cell line alternatives

Frozen cells are supplied in a vial containing 1 mL of cells at 1 x 107 viable cells/mL in Expi293 Expression Medium and 10% DMSO. The cells should be thawed directly into Expi293 Expression Medium

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 1 vial Expi293F GnTI- Cells (1x107 cells/vial), store in vapor-phase liquid nitrogen

Product Line Expi293
Quantity 1 vial
Shipping Condition Dry Ice
Can I leave my Expi293F culture expressing for more than 7 days?

The growth and expression characteristics of Expi293F cells are such that, by 7 days post-transfection, the culture medium should be close to being spent and maximal protein expression should have already been achieved. Continued incubation will result in a large decrease in cell culture viability.

If I let my Expi293F cells grow to a higher density than recommended, will they be okay?

While the Expi293 Expression Medium can support much higher cell densities, we do not recommend growing your Expi293F cultures beyond 5-6 x 10e6 cells/mL, as subsequent transfection and protein expression efficiencies may be reduced. At higher densities, there is also the increased possibility of reaching the point of reduced culture viability. If your seed culture does exceed 5-6 x 10e6 cells/mL, passage them once or twice as detailed in the manual, monitoring them for viability and growth rate. Perform a test transfection using the Protein Expression Control IgG or an expression construct of known yield to determine if cell expression performance has been impacted.

What method do you recommend for quantifying rabbit IgG expression from pRABBIT IgG IRES-EmGFP Positive Control Vector, Cat. No. A39243?

The formation of intact IgG molecules may be quantified using a sandwich ELISA designed to capture and detect rabbit IgG. Besides the rabbit IgG positive control vector, reagents, and consumables that are included in the kit, you will also need purified rabbit IgG to be used as a standard, F(ab')2-Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, HRP (Cat. No. A10547), Protein A Coated Plates, Clear, 96-Well (Cat. No. 15130), TMB Substrate Kit (Cat. No. 34021), SuperBlock (TBS) Blocking Buffer (Cat. No. 37535), and PBS or TBS buffer for washes. There is an example procedure in our Protein A Coated Plates manual (https://tools.thermofisher.com/content/sfs/manuals/MAN0011310_Thermo Scientific_ProteinA_G_AG_Coat_96Well_UG.pdf). Please note that our R&D scientists determine titer values from crude cell culture supernatants using a Pall Life Sciences FortéBio Octet instrument equipped with a Protein A biosensor.

Do you offer a GFP-expressing mammalian expression vector that I can use as a control to monitor my transfection and expression with the Expi systems?

We offer pRABBIT IgG IRES-EmGFP Positive Control Vector, Cat. No. A39243, which you can use to monitor your transfection and expression.

How do I culture Expi293F GnTI-, Expi293F Inducible, or Expi293F Inducible GnTI- cells?

Expi293F GnTI-, Expi293F Inducible, and Expi293F Inducible GnTI- cell lines were derived from the parental Expi293F Cells (Cat. No. A14527) and therefore, share similar morphology and growth characteristics. They are cultured in Expi293 Expression Medium and transfected using ExpiFectamine 293 Transfection Kit following our standard Expi293 protocol.

How soon after thawing can I use my Expi293F cells and for how long are they good to use?

Expi293F cells must be recovered from freezing and passaged at least 3 times using the procedure outlined in the manual to ensure optimal performance. Cells should maintain performance for at least 30 passages if maintained in accordance with the protocol in the manual.


WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.