Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ ExpiCHO™ Expression System Kit

Catalog No. p-200089243
Encompass_Preferred
Change view
Click to view available options
Cell Line:
CHO-S, SFM Adapted
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cell Line
A29133 CHO-S, SFM Adapted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A29133 Supplier Gibco™ Supplier No. A29133
Only null left
Add to Cart
Add to Cart

Includes

2 vials frozen ExpiCHO-S™ cells, 1 bottle 1000mL ExpiCHO™ Expression Medium, 1 ExpiFectamine™ CHO transfection kit for 1 L of culture, 1 vial Antibody Expressing Positive Control Vector, 1 bottle 100mL OptiPRO™ SFM; store all components at +2° - +8° C, except frozen cells stored in liquid nitrogen

The ExpiCHO Expression System Kit combines the power of rapid, ultra-high-yield transient protein production in suspension culture with the benefit of expression in Chinese hamster ovary (CHO) cells.

The ExpiCHO Expression System Kit combines the power of rapid, ultra–high-yield transient protein production in suspension culture with the benefit of expression in Chinese hamster ovary (CHO) cells. It is based on high-density culture of ExpiCHO-S cells in ExpiCHO Expression Medium. Transient expression is powered by ExpiFectamine CHO transfection reagent, designed specifically for high-density CHO suspension culture, in combination with a specialized transfection enhancer and culture feed.

All components work together synergistically with expression yields of 1–3 g per L of transfected culture demonstrated for some antibody and non-antibody proteins.

The ExpiCHO Expression System enables:

  • Milligram to multigram yields of recombinant protein per liter of culture in 5–14 days post-transfection
  • Robust and reproducible transfection results, for greater confidence in results and ability to use CHO cells from research to discovery
  • Transfection efficiency of > 90% for high-density suspension CHO cell culture, with matching transfection enhancer and culture feed to boost transfection performance and protein expression
  • Use of one-third to one-half less plasmid DNA than other transfection reagents 
  • Flexible, reliable culture and transfection protocols with multiple options that readily fit into current transient expression workflows
  • Scalable expression for culture formats from  <1 mL in deep well plates up to 5 L when using Nalgene angled-bottom shake flasks

The components included in the ExpiCHO Expression System Kit are: 2 vials of frozen ExpiCHO-S cells, 1 L of ExpiCHO Expression Medium, 1 ExpiFectamine CHO Transfection Kit sufficient to transfect 1 L of culture, OptiPRO SFM, and an antibody-expressing positive control vector.

ExpiCHO-S cells, ExpiCHO Expression Medium, ExpiFectamine CHO Transfection Kit, and OptiPRO SFM may also be purchased separately.

All components of the system are animal origin–free.

TRUSTED_SUSTAINABILITY

Specifications

Type Expression System
Protein Tag Untagged
Delivery Type Transfection
Quantity 1 kit
Content And Storage • 2 vials frozen ExpiCHO-S cells
• 1000 mL ExpiCHO Expression Medium
• 1 ExpiFectamine CHO Transfection Kit for 1 L of culture
• 1 vial Antibody Expressing Positive Control Vector
• 100 mL OptiPRO SFM

Store all components at 2-8°C, except frozen cells which should be stored in liquid nitrogen.
Shipping Condition Approved for shipment at Room Temperature or on Wet or Dry Ice
Cell Line CHO-S, SFM Adapted
Expression System Mammalian
Product Line ExpiCHO
For Use With (Application) Protein Production, Constitutive Expression
Promoter CMV
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.