Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ ExpiFectamine™ 293 Met (–) Transfection Kit

Catalog No. A39249
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A39249 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A39249 Supplier Gibco™ Supplier No. A39249
Get 15% Off Cell Culture Products
Use promo code CELLS15
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

ExpiFectamine 293 Met (-) Transfection Kit is a core component of the Expi293 Protein Labeling Kit for rapid, high-yield transient protein expression from Expi293F cells for applications where labelling of methionine residues in proteins is desired

Kit components are specifically formulated without methionine, allowing users to add labeled methionine of their choice.

Featues:

  • Methionine-free formulation
  • 2- to 10-fold higher protein yields compared to other transfection reagents used for high-density 293 cell culture
  • Same transient transfection protocol as ExpiFectamine 293 Transfection Kit
  • Scalable transfection from small to large scale while maintaining equivalent volumetric protein yields

Specifically designed to work with Expi293F cell lines as part of the Expi293 Expression System

Components included in the ExpiFectamine 293 Met (–) Transfection Kit are:

  • 2.7 mL ExpiFectamine 293 Reagent
  • 5 mL ExpiFectamine 293 Transfection Enhancer
  • 1 x 50 mL ExpiFectamine 293 Met (–) Transfection Enhancer 2
  • 100 mL Opti-Plex Complexation Buffer (methionine-free)

Component volumes are sufficient to transfect 1 L of culture

TRUSTED_SUSTAINABILITY

Specifications

Classification AOF, Chemically Defined, Methionine-Free, Protein-Free, Serum Free
Content And Storage • 3 x 0.9 mL ExpiFectamine 293 Transfection Reagent, store at 2–8°C
• 5 mL ExpiFectamine 293 Enhancer 1, store at 2–8°C
• 50 mL ExpiFectamine 293 Met (–) Enhancer 2, store at 2–8°C
• 100 mL Opti-Plex Complexation Buffer, store at 2–8°C (protect from light)

Cell Type Established Cell Lines
For Use With (Application) Transfection
Product Line ExpiFectamine
Product Type Transfection Kit
Quantity 1 Kit
Shipping Condition Approved for shipment at Room Temperature or on Wet or Dry Ice
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.