Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Minuteman Explosion Proof Vacuum Kit, Wet/Dry, 30H, 1 ea

Catalog No. 502867581
Encompass

This explosion-proof vacuum tool kit is designed for use with 30H series vacuums, providing essential accessories for both wet and dry material collection in hazardous environments.

  • Squeegee, 14 in
  • Round dusting brush, 5 in
  • Hose assembly, 12 ft x 1 1/2 in
  • Curved pipe tool, 6 ft x 1 1/2 in
  • Gulper tool, 12 in
  • Fiber floor tool, 14 in

Catalog No. 50-286-7581 Supplier MSC Supplier No. 57122376
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This explosion-proof vacuum tool kit is designed for use with 30H series vacuums, providing essential accessories for both wet and dry material collection in hazardous environments.

  • Squeegee, 14 in
  • Round dusting brush, 5 in
  • Hose assembly, 12 ft x 1 1/2 in
  • Curved pipe tool, 6 ft x 1 1/2 in
  • Gulper tool, 12 in
  • Fiber floor tool, 14 in

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.