Learn More
Cole-Parmer KNF UN726.3FTI EX Explosion Proof Motor Vacuum Pump, PTFE/PTFE; 0.6 cfm/29.5"Hg
Catalog No. NC1517016 Shop All Cole-Parmer Products
Encompass
Diaphragm pumps are fitted with Class I, Division 1, Groups C & D explosion-proof motors. Designed for continuous, heavy-duty use, ball bearings are used throughout to provide exceptional reliability. Models are fitted with a pre-wired connection box and are tested and ready to install into your system. Contact one of our Application Specialists for assistance with selecting the proper pump for your needs.
Catalog No. NC1517016 Supplier Cole-Parmer Supplier No. 0705763
Diaphragm pumps are fitted with Class I, Division 1, Groups C & D explosion-proof motors. Designed for continuous, heavy-duty use, ball bearings are used throughout to provide exceptional reliability. Models are fitted with a pre-wired connection box and are tested and ready to install into your system. Contact one of our Application Specialists for assistance with selecting the proper pump for your needs.