Learn More
Pipette Supplies, Inc. Xplorer / Xplorer Plus Lithium-Polymer Rechargeable Battery (Eppendorf)
SDP
Catalog No. NC1220130
Encompass
Eppendorf OEM Lithium-Polymer Rechargeable Battery.3.7v, 750mAh, 2.775WhBattery must be charged prior to use.Please note that Pipette Supplies only ships this battery via Ground Shipping.Made in China
Catalog No. NC1220130 Supplier Pipette Supplies, Inc. Supplier No. EP 4861601000
Eppendorf OEM Lithium-Polymer Rechargeable Battery.3.7v, 750mAh, 2.775WhBattery must be charged prior to use.Please note that Pipette Supplies only ships this battery via Ground Shipping.Made in China