Learn More
Med Vet International Explorer/Measuring Probe, 6.5in, J0042Q
Catalog No. 5000585381
Encompass_Preferred
The delicate explorer tip is used for increased tactical sensitivity in the gingival sulcus. It locates subgingival calculus and can be used to check tooth mobility. The periodontal measuring probe is graduated in millimeters. It is held parallel to the long axis of the tooth and measures gingival sulcus depth. 6 1/2".
Catalog No. 50-005-85381 Supplier Med Vet International Supplier No. JOR42Q
The delicate explorer tip is used for increased tactical sensitivity in the gingival sulcus. It locates subgingival calculus and can be used to check tooth mobility. The periodontal measuring probe is graduated in millimeters. It is held parallel to the long axis of the tooth and measures gingival sulcus depth. 6 1/2".