Learn More
Med Vet International Explorer/Measuring Probes - Explorer/Measuring Probe Standard - Silicone, J0042QS, Each
Catalog No. 5000585382
Encompass_Preferred
The delicate explorer tip is used for increased tactical sensitivity in the gingival sulcus. It locates subgingival calculus and can be used to check tooth mobility. The periodontal measuring probe is graduated in millimeters. It is held parallel to the long axis of the tooth and measures gingival sulcus depth.
Catalog No. 50-005-85382 Supplier Med Vet International Supplier No. JOR42QS
The delicate explorer tip is used for increased tactical sensitivity in the gingival sulcus. It locates subgingival calculus and can be used to check tooth mobility. The periodontal measuring probe is graduated in millimeters. It is held parallel to the long axis of the tooth and measures gingival sulcus depth.