Learn More
Chemglass Life Sciences 1/4 HP DC C1D2 90VDC Permanent Magnet Explosion Proof Motor Only. Complete with 5:1 Gearbox and Stand Mounting Adapter.
SDP
Catalog No. 50401820
Encompass_Preferred
Explosion Proof Motor Only. Explosion proof (XP) motor features a 5:1 right angle gearbox to reduce the overall height above support frame to 10.5¿. Motor is supplied with a 30ft 18/3 SJEOO cord with twist lock connector. XP motor must be used in conjunction with our digital motor controller, and mounts to all our support frames via the supplied mounting bracket. PLEASE NOTE: A LICENSED ELECTRICIAN SHOULD BE CONSULTED FOR ALL FINAL CONNECTIONS. DIGITAL MOTOR CONTROLLER IS NOT EXPLOSION PROOF AND MUST BE MOUNTED OUTSIDE THE HOOD/XP ENVIRONMENT.
Catalog No. 50-401-820 Supplier Chemglass Life Sciences Supplier No. CG2033X11
Explosion Proof Motor Only. Explosion proof (XP) motor features a 5:1 right angle gearbox to reduce the overall height above support frame to 10.5¿. Motor is supplied with a 30ft 18/3 SJEOO cord with twist lock connector. XP motor must be used in conjunction with our digital motor controller, and mounts to all our support frames via the supplied mounting bracket. PLEASE NOTE: A LICENSED ELECTRICIAN SHOULD BE CONSULTED FOR ALL FINAL CONNECTIONS. DIGITAL MOTOR CONTROLLER IS NOT EXPLOSION PROOF AND MUST BE MOUNTED OUTSIDE THE HOOD/XP ENVIRONMENT.