Learn More
Grainger Explosionproof Box Gray
Catalog No. 502766333
Encompass_Preferred
The Grainger weatherproof electrical boxes are made of durable polycarbonate plastic and feature a gasketed cover that seals out moisture, dust and insects. They are UL listed for wet locations and come in a variety of sizes to accommodate different wire and conduit sizes. The clear covers allow easy inspection of wiring. These boxes can be surface or flush mounted for indoor or outdoor electrical applications that require protection from the elements.
Catalog No. 50-276-6333 Supplier Grainger Supplier No. 16X410
The Grainger weatherproof electrical boxes are made of durable polycarbonate plastic and feature a gasketed cover that seals out moisture, dust and insects. They are UL listed for wet locations and come in a variety of sizes to accommodate different wire and conduit sizes. The clear covers allow easy inspection of wiring. These boxes can be surface or flush mounted for indoor or outdoor electrical applications that require protection from the elements.