Learn More
Medicus Health EXPANDABLE TUBE RACKS, POLYPROPYLENE, 10-13MM TUBES, 2 PK
SDP
Catalog No. NC1929591
Encompass
∙ Exclusive Expandable Tube Racks, Polypropylene for 10-13mm Tubes allow you to combine racks together with an easy positive "click stop" to make your tube capacity as large as you need it ∙You can even combine multiple tube sizes into one tube racks to increase your capacity∙These patented tube racks lock together so there is no sliding apart so they are easy to carry and store∙To disassemble, simply slide the racks apart - it's that easy∙Durable polypropylene racks are autoclavable tube racks and easy to assemble∙Tube Racks will not break or become brittle in low temperature refrigerators∙Tube Racks are easy to clean and disinfect∙Each tube rack has 50 places in two 25 hole configurations for 10-13mm diameter tubes∙Includes molded-in index coordinates and flat areas for labeling∙Each tube rack measures: 8.75"L x 3.75"W x 5.25"H
Catalog No. NC1929591 Supplier Medicus Health Supplier No. 2288M2
∙ Exclusive Expandable Tube Racks, Polypropylene for 10-13mm Tubes allow you to combine racks together with an easy positive "click stop" to make your tube capacity as large as you need it ∙You can even combine multiple tube sizes into one tube racks to increase your capacity∙These patented tube racks lock together so there is no sliding apart so they are easy to carry and store∙To disassemble, simply slide the racks apart - it's that easy∙Durable polypropylene racks are autoclavable tube racks and easy to assemble∙Tube Racks will not break or become brittle in low temperature refrigerators∙Tube Racks are easy to clean and disinfect∙Each tube rack has 50 places in two 25 hole configurations for 10-13mm diameter tubes∙Includes molded-in index coordinates and flat areas for labeling∙Each tube rack measures: 8.75"L x 3.75"W x 5.25"H