Learn More
Grainger Exposed Manual Flush Valve Top Spud
Catalog No. 502768458
Encompass_Preferred
The American Standard 16T985 is an automatic flush valve for toilets. It features a chrome plated body with a polished brass shank and adjustable tailpiece. The flush valve has a diaphragm type flushometer that provides automatic flushing when objects pass its sensor. It requires a 1" top spud connection and has an exposed style actuator with vandal resistant stop cap. The valve is ADA compliant.
Catalog No. 50-276-8458 Supplier Grainger Supplier No. 16T985
The American Standard 16T985 is an automatic flush valve for toilets. It features a chrome plated body with a polished brass shank and adjustable tailpiece. The flush valve has a diaphragm type flushometer that provides automatic flushing when objects pass its sensor. It requires a 1" top spud connection and has an exposed style actuator with vandal resistant stop cap. The valve is ADA compliant.