Learn More
Grainger Exposed Top Spud Automatic Flush Valve
Catalog No. 502766939
Encompass_Preferred
The Zurn 307A13 is an exposed flush valve for toilets and urinals. It is made of chrome plated brass and has a 1-1/4" top spud inlet. This valve features a vandal resistant stop cap and sweat solder kit. It has an adjustable tailpiece that fits 2" or 3" connections. The flush valve is designed for manual flush only and has a float with chloramine resistant components. This valve meets ASSE 1037 and CSA B125 standards and is WaterSense certified. The Zurn 307A13 exposed flush valve is made in the USA.
Catalog No. 50-276-6939 Supplier Grainger Supplier No. 16T987
The Zurn 307A13 is an exposed flush valve for toilets and urinals. It is made of chrome plated brass and has a 1-1/4" top spud inlet. This valve features a vandal resistant stop cap and sweat solder kit. It has an adjustable tailpiece that fits 2" or 3" connections. The flush valve is designed for manual flush only and has a float with chloramine resistant components. This valve meets ASSE 1037 and CSA B125 standards and is WaterSense certified. The Zurn 307A13 exposed flush valve is made in the USA.