Learn More
Cryoguard Corporation EXPOSER INDICTOR 10/BOX
SDP
Catalog No. NC0556793
Encompass
Cryoguard Thermal Exposure Indicator •Signal temperature: -100°C •Activated indicator turns red when exposed to temperatures at or above -100°C. •Used for monitoring products shipped or stored in liquid nitrogen that need to be kept at or below -100°C.•Popular for monitoring stem cells & more. •Recommended for monitoring environments at or below -125°C •Cryoguard indicators have a shelf life of eight months. The shelf life applies whether the indicator is activated or unactivated. An expiration date appears on the label on each indicator.
Catalog No. NC0556793 Supplier Cryoguard Corporation Supplier No. M10010
Cryoguard Thermal Exposure Indicator •Signal temperature: -100°C •Activated indicator turns red when exposed to temperatures at or above -100°C. •Used for monitoring products shipped or stored in liquid nitrogen that need to be kept at or below -100°C.•Popular for monitoring stem cells & more. •Recommended for monitoring environments at or below -125°C •Cryoguard indicators have a shelf life of eight months. The shelf life applies whether the indicator is activated or unactivated. An expiration date appears on the label on each indicator.