Learn More
Expression Systems Expres2 TR (Transfection Reagent), 75 ul (included in BestBac kits)
SDP
Catalog No. NC0625261
Encompass_Preferred
In collaboration with Expres2ion Biotechnologies, Expression Systems offers Expres2 TR Transfection Reagent, formulated and optimized for insect cell culture. Expres2 TR is a cationic lipid formulation designed for the co-transfection or transfection step in baculovirus vector production.
• Formulation works seamlessly with Transfection Medium
• Optimized for use in insect cell culture
• Consistently high transfection rates Increases virus vector production and transient expression
• In stock for immediate shipment
To download Instructions for Use and/or COA please visit: www.expressionsystems.com
Catalog No. NC0625261 Supplier Expression Systems Supplier No. 95055075
In collaboration with Expres2ion Biotechnologies, Expression Systems offers Expres2 TR Transfection Reagent, formulated and optimized for insect cell culture. Expres2 TR is a cationic lipid formulation designed for the co-transfection or transfection step in baculovirus vector production.
• Formulation works seamlessly with Transfection Medium
• Optimized for use in insect cell culture
• Consistently high transfection rates Increases virus vector production and transient expression
• In stock for immediate shipment
To download Instructions for Use and/or COA please visit: www.expressionsystems.com