Learn More
Bioworld Express Coomassie Stain, R-250, 100 mL
Catalog No. 50255835
Encompass_Preferred
Express Coomassie Stain detects as little as 15 ng of protein and gives complete band detection within 50 minutes. Its ideal for densitometry, and its able to be restained with silver. This stain can be used more than once. Express Coomassie is a colloidal suspension of Coomassie blue that preferentially binds to protein and eliminates the unwanted destaining step.Appearance: LiquidHS Code : 32041200Melting Point: 100 °C
Catalog No. 50-255-835 Supplier Bioworld Supplier No. 403400241
Express Coomassie Stain detects as little as 15 ng of protein and gives complete band detection within 50 minutes. Its ideal for densitometry, and its able to be restained with silver. This stain can be used more than once. Express Coomassie is a colloidal suspension of Coomassie blue that preferentially binds to protein and eliminates the unwanted destaining step.Appearance: LiquidHS Code : 32041200Melting Point: 100 °C