Learn More
Takara Bio EXPRESS HYP HYBRIDIZATION
SDP
Catalog No. NC9747391
Encompass
ExpressHyb Hybridization Solution, 1 L, Faster hybridizations with increased sensitivity, Ideal for cDNA array, Northern blot and Southern blot, and colony hybridization, Compatible with radioactive or nonisotopic detection systems
Catalog No. NC9747391 Supplier Takara Bio Supplier No. 636833
Add to Cart
Add to Cart
ExpressHyb Hybridization Solution, 1 L, Faster hybridizations with increased sensitivity, Ideal for cDNA array, Northern blot and Southern blot, and colony hybridization, Compatible with radioactive or nonisotopic detection systems