Learn More
Genscript Corporation ExpressPlus™ PAGE Gel,10×8, 4-20%, 12 wells
Supplier:  Genscript Corporation M42012
Encompass
GenScript ExpressPlus™ PAGE Gels are high-performance precast mini polyacrylamide gels specially designed for large loading volumes. The unique design of the cassette gives better band resolution and significantly improves the sample distribution in the loading wells which increases the evenness of the band. ExpressPlus™ PAGE Gels are casted in a weak acidic pH buffer that minimizes the hydrolysis of polyacrylamide and results in extra gel stability.Manufactured without SDS, ExpressPlus™ PAGE Gels are ideal for SDS-PAGE electrophoresis depending on the running buffer and transfer buffer used. The proprietary gel-casting techniques provide excellent batch-to-batch consistency and guarantee a reliable migration pattern. Using specially formulated Tris-MOPS running buffer, ExpressPlus™ PAGE Gels enable proteins to be separated quickly and easily for subsequent detection by staining or western blotting.
Catalog No. NC0813248