Learn More
Genscript Corporation ExpressPlus PAGE Gel 10x8 4
Catalog No. 5000058517
Encompass
Bis-tris 4-12% 10 well br Cassette size 100 x 84 5 x 4 7 mm (L x W x T) br Gel size 80 x 70 x 1 mm (L x W x T)
Catalog No. 50-000-58517 Supplier Genscript Corporation Supplier No. M41210
Bis-tris 4-12% 10 well br Cassette size 100 x 84 5 x 4 7 mm (L x W x T) br Gel size 80 x 70 x 1 mm (L x W x T)