Learn More
Genscript Corporation ExpressPlus™ PAGE Gel,10x8, 4-12%, 15 wells
Catalog No. NC1486979
Encompass
GenScript ExpressPlus™ PAGE Gels are high-performance precast mini polyacrylamide gels specially designed for large loading volumes. The unique design of the cassette gives better band resolution and significantly improves the sample distribution in the loading wells which increases the evenness of the band. ExpressPlus™ PAGE Gels are casted in a weak acidic pH buffer that minimizes the hydrolysis of polyacrylamide and results in extra gel stability.
Catalog No. NC1486979 Supplier Genscript Corporation Supplier No. M41215
GenScript ExpressPlus™ PAGE Gels are high-performance precast mini polyacrylamide gels specially designed for large loading volumes. The unique design of the cassette gives better band resolution and significantly improves the sample distribution in the loading wells which increases the evenness of the band. ExpressPlus™ PAGE Gels are casted in a weak acidic pH buffer that minimizes the hydrolysis of polyacrylamide and results in extra gel stability.