Learn More
Cayman Chemical Extracellular Vesicle Isolation Kit (plasma and serum), 1 ea
SDP
Catalog No. 502893546
Encompass_Preferred
Cayman’s Extracellular Vesicle Isolation Kit (plasma and serum) provides a simple, quick, and convenient precipitation-based method for isolating extracellular vesicles from serum and plasma. The volume of Extracellular Vesicle Isolation Reagent provided is sufficient for processing up to 24 ml of serum or plasma, and it enables rapid isolation within one hour. Also provided is a storage medium for stabilizing extracellular vesicles during frozen storage. This optional reagent will not interfere with protein assays and is formulated to be compatible with typical downstream analyses.
Catalog No. 50-289-3546 Supplier Cayman Chemical Supplier No. 7024201 EA
Cayman’s Extracellular Vesicle Isolation Kit (plasma and serum) provides a simple, quick, and convenient precipitation-based method for isolating extracellular vesicles from serum and plasma. The volume of Extracellular Vesicle Isolation Reagent provided is sufficient for processing up to 24 ml of serum or plasma, and it enables rapid isolation within one hour. Also provided is a storage medium for stabilizing extracellular vesicles during frozen storage. This optional reagent will not interfere with protein assays and is formulated to be compatible with typical downstream analyses.