Learn More
Bioworld EXS III ™ Basal Medium, 25 L
Catalog No. 50254903
Encompass_Preferred
EXS III™ Basal MediumContains Micronutrients, Macronutrients, myo-Inositol, and ThiamineSucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 25L, or 50L of mediumPlant tissue culture testedFor the culture of foliage plants such as Syngonium, Ficus, and SpathiphyllumWorking Concentration: 3.87 g/LpH: 4.25–5.25Soluble in water
Catalog No. 50-254-903 Supplier Bioworld Supplier No. 306301641
EXS III™ Basal MediumContains Micronutrients, Macronutrients, myo-Inositol, and ThiamineSucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 25L, or 50L of mediumPlant tissue culture testedFor the culture of foliage plants such as Syngonium, Ficus, and SpathiphyllumWorking Concentration: 3.87 g/LpH: 4.25–5.25Soluble in water