Learn More
Bioworld EXS III ™ Basal Salt Medium, w/ Adenine, 50 L
Catalog No. 50254902
Encompass_Preferred
EXS IIIa™ Basal Salt Medium w/ AdenineContains Micrnutrients, Macronutrients, myo-Inositol, Thiamine HCl, and AdenineSucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 25L or 50L of mediumPlant tissue culture testedFor the culture of foliage plants such as Syngonium, Ficus, and SpathiphyllumWorking Concentration: 3.95 g/LpH: 3.5–4.5Soluble in water
Catalog No. 50-254-902 Supplier Bioworld Supplier No. 306301632
EXS IIIa™ Basal Salt Medium w/ AdenineContains Micrnutrients, Macronutrients, myo-Inositol, Thiamine HCl, and AdenineSucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 25L or 50L of mediumPlant tissue culture testedFor the culture of foliage plants such as Syngonium, Ficus, and SpathiphyllumWorking Concentration: 3.95 g/LpH: 3.5–4.5Soluble in water