Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tripak Buffer Pouches, FLIR Systems
SCI_ED

Catalog No. S66292
Encompass
Encompass_Preferred
Change view
Click to view available options
For Use With (Application):
pH Measurement
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
S66292 pH Measurement
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S66292 Supplier GENIE GROUP Supplier No. PH103
May include imposed supplier surcharges.
Add to Cart
Add to Cart
  • Easy to use, already made fresh pH buffers for calibration
  • 6ea. of 4, 7, 10pH plus 2 Rinsing solutions
  • Helpful table printed on the pouch shows what the pH value should be vs. the temperature of the buffer solution measured
  • Rinsing solution for cleaning the electrode before taking the next measurement to prevent contamination
  • Convenient pouch eliminates the need for a cup to store buffer or rinsing solution

Specifications

Chemical Name or Material Tripak Buffer Pouches
For Use With (Application) pH Measurement
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.