Learn More
Chemglass Life Sciences 10ft Extension Cord, Type "J" (Black), Standard to Mini Plug
SDP
Supplier:  Chemglass Life Sciences CG349901
Encompass_Preferred
10ft Extension Cord, Type "J" (Black), Standard to Mini Plug. Coiled extension cords to connect thermocouples to instrumentation such as temperature monitors, controllers and data loggers. Cords are 10ft long and made of 26-gauge multistrand wire with PVC insulation. All cords have a female standard (OST) with a choice of a male standard (OST) or mini (SMP) connection.
Catalog No. CG349901