Learn More
Bellco Glass, Inc. Ext. Heat/Cool Coil, 36L
SDP
Catalog No. 501944801
Encompass_Preferred
Heating and cooling coils are used for cell culture temperature manipulation within the bioreactor vessel. Must be hooked up to a water circulating mechanism for heating or cooling. MADE IN THE USA. 316l electropolished stainless steel. OD X OAL: 3 1/4 X 22". #TURNS: 5 1/2. FITS 100mm CAP ASSY OR LARGE FLANGE HEADPLATE
Catalog No. 50-194-4801 Supplier Bellco Glass, Inc. Supplier No. 196450136
Heating and cooling coils are used for cell culture temperature manipulation within the bioreactor vessel. Must be hooked up to a water circulating mechanism for heating or cooling. MADE IN THE USA. 316l electropolished stainless steel. OD X OAL: 3 1/4 X 22". #TURNS: 5 1/2. FITS 100mm CAP ASSY OR LARGE FLANGE HEADPLATE