Learn More
SAI Infusion Technologies Micro Flow Extension Line - 36", Male & Female Luers
SDP
Catalog No. NC0476058
Encompass
36" Micro Flow Extension line with male and female luer adapters, 287ul dead volume. SAI extension lines are ideal for connecting between pump and swivel or pump and catheter in animal infusion studies. Made with non-DEHP PVC tubing and available in a variety of standard lengths. Custom sizes are also available. 50/pk. Product is for laboratory animal research only.
Catalog No. NC0476058 Supplier SAI Infusion Technologies Supplier No. EXT36
36" Micro Flow Extension line with male and female luer adapters, 287ul dead volume. SAI extension lines are ideal for connecting between pump and swivel or pump and catheter in animal infusion studies. Made with non-DEHP PVC tubing and available in a variety of standard lengths. Custom sizes are also available. 50/pk. Product is for laboratory animal research only.