Learn More
MSC Fantech Duct Fan, Galvanized Steel, 116 CFM, 115 V, 1 ea
Catalog No. 5000509381
Encompass
This direct drive centrifugal fan is designed for residential and light commercial ventilation applications requiring exterior mounting. The unit features a galvanized steel housing with a baked powder-coat finish and is suitable for installation on pitched roofs or walls. It utilizes an external rotor motor with permanently lubricated sealed ball bearings to provide efficient, long-term operation.
  • Features a weather-resistant design for exterior roof or wall installation
  • Equipped with built-in thermal overload protection and automatic reset
  • Includes an integrated motorized impeller for vibration-free and quiet operation
  • Offers 100% speed controllability for customized airflow
  • Capable of operating in airstream temperatures up to 60°C (140°F)
  • Includes a 5-year warranty
Catalog No. 50-005-09381 Supplier MSC Supplier No. 72096910
This direct drive centrifugal fan is designed for residential and light commercial ventilation applications requiring exterior mounting. The unit features a galvanized steel housing with a baked powder-coat finish and is suitable for installation on pitched roofs or walls. It utilizes an external rotor motor with permanently lubricated sealed ball bearings to provide efficient, long-term operation.
  • Features a weather-resistant design for exterior roof or wall installation
  • Equipped with built-in thermal overload protection and automatic reset
  • Includes an integrated motorized impeller for vibration-free and quiet operation
  • Offers 100% speed controllability for customized airflow
  • Capable of operating in airstream temperatures up to 60°C (140°F)
  • Includes a 5-year warranty