Learn More
BECTON DICKINSON INFUSION THERAPY Extension Set MaxZero, 50/cs
Catalog No. NC1836508
Encompass_Preferred
Needle-Free Connectors, Set Length 18 cm, Priming Volume 0.5 ml, and Bore 1.5 mm.
This item is not returnable. View return policy
Catalog No. NC1836508 Supplier BECTON DICKINSON INFUSION THERAPY Supplier No. MZ5303
Needle-Free Connectors, Set Length 18 cm, Priming Volume 0.5 ml, and Bore 1.5 mm.