Learn More
Andwin Scientific EXT SET W/MALE LUER LOC CS/48
SDP
Catalog No. NC9776295
Encompass_Preferred
The Baxter extension set with male luer lock adapter has an approximate length of 30 inches and an approximate volume of 4.0 mL. This product is sold in a case of 48.
Catalog No. NC9776295 Supplier Andwin Scientific Supplier No. 2C5645
The Baxter extension set with male luer lock adapter has an approximate length of 30 inches and an approximate volume of 4.0 mL. This product is sold in a case of 48.