Learn More
Grainger M-Struts Extension Strut, Mechanical Traction, Carbon Steel, 12 in. Stroke
Catalog No. 502779432
Encompass_Preferred
This mechanical traction extension strut is designed for precise force management and load support. Constructed from carbon steel with a durable nitride finish, this component provides reliable, fixed resistance for demanding industrial applications. The strut operates effectively at temperatures up to 400°F.
  • Provides an extension force of 30 lbf
  • Features a 12 in. stroke length
  • Includes a 0.315 in. diameter rod and 0.71 in. diameter cylinder
  • Constructed with an M6 x 1 rod thread size
  • Maintains fixed resistance in an extension-only damper configuration
  • FDA approved for specialized usage requirements
Catalog No. 50-277-9432 Supplier Grainger Supplier No. 19MU01
This mechanical traction extension strut is designed for precise force management and load support. Constructed from carbon steel with a durable nitride finish, this component provides reliable, fixed resistance for demanding industrial applications. The strut operates effectively at temperatures up to 400°F.
  • Provides an extension force of 30 lbf
  • Features a 12 in. stroke length
  • Includes a 0.315 in. diameter rod and 0.71 in. diameter cylinder
  • Constructed with an M6 x 1 rod thread size
  • Maintains fixed resistance in an extension-only damper configuration
  • FDA approved for specialized usage requirements