Learn More
Grainger M-STRUTS Mechanical Traction Extension Strut, Carbon Steel, 8 in. Stroke
Catalog No. 502779319
Encompass_Preferred
This mechanical gas spring features a durable wire compression spring design, making it ideal for applications in harsh, dirty, or dusty environments. Unlike conventional gas-filled springs, this mechanical alternative maintains a regular linear force curve without damping at full extension. Constructed from carbon steel, this unit is engineered to withstand impact, vibration, and prolonged static loads, making it suitable for infrequently moved structures like emergency exits.
  • Features a robust carbon steel construction.
  • Provides a consistent linear force curve.
  • Operates in a wide temperature range of -40°C to +200°C.
  • Compatible with standard threaded end fittings for flexible installation.
  • Designed for durability in environments prone to vibration and shock.
Catalog No. 50-277-9319 Supplier Grainger Supplier No. 19MT80
This mechanical gas spring features a durable wire compression spring design, making it ideal for applications in harsh, dirty, or dusty environments. Unlike conventional gas-filled springs, this mechanical alternative maintains a regular linear force curve without damping at full extension. Constructed from carbon steel, this unit is engineered to withstand impact, vibration, and prolonged static loads, making it suitable for infrequently moved structures like emergency exits.
  • Features a robust carbon steel construction.
  • Provides a consistent linear force curve.
  • Operates in a wide temperature range of -40°C to +200°C.
  • Compatible with standard threaded end fittings for flexible installation.
  • Designed for durability in environments prone to vibration and shock.