Learn More
Grainger M-STRUTS Gas Spring, Carbon Steel, 12 in Stroke, 105 lbf, 1 ea
Catalog No. 502783240
Encompass_Preferred
This gas spring provides controlled force to assist in opening or closing components such as windows, seats, covers, and hatchbacks. Designed for mechanical traction, it utilizes compressed gas and internal liquid to facilitate smooth movement. It is constructed from durable carbon steel with a nitride finish and features a fixed resistance.
  • Extended length of 36-13/16 in.
  • Compressed length of 24-13/16 in.
  • Maximum operating temperature of 400 °F
  • Thread size of M10x1.5
  • Stroke length of 12 in.
  • Force of 105 lbf
  • Cylinder diameter of 1.1 in. and rod diameter of 0.55 in.
  • End fittings not included
Catalog No. 50-278-3240 Supplier Grainger Supplier No. 19MU05
This gas spring provides controlled force to assist in opening or closing components such as windows, seats, covers, and hatchbacks. Designed for mechanical traction, it utilizes compressed gas and internal liquid to facilitate smooth movement. It is constructed from durable carbon steel with a nitride finish and features a fixed resistance.
  • Extended length of 36-13/16 in.
  • Compressed length of 24-13/16 in.
  • Maximum operating temperature of 400 °F
  • Thread size of M10x1.5
  • Stroke length of 12 in.
  • Force of 105 lbf
  • Cylinder diameter of 1.1 in. and rod diameter of 0.55 in.
  • End fittings not included