Learn More
Grainger M-Struts Extension Strut, Carbon Steel, 10 in. Stroke, 21 lbf., 1 ea
Catalog No. 502779219
Encompass_Preferred
The mechanical traction extension strut is designed for precise load bearing and controlled extension. Engineered from carbon steel with a nitride finish, this robust component ensures durability and reliability in various mechanical systems. It operates as an extension-only damper with a fixed resistance design, providing stable performance across a temperature range up to 400 °F.
  • Stroke length of 10 in.
  • Constructed from carbon steel with a nitride finish
  • Fixed resistance mechanism for consistent operation
  • Maximum operating temperature of 400 °F
  • Features an extended length of 30 19/64 in. and a compressed length of 20 19/64 in.
  • Equipped with an M6 x 1 thread size
Catalog No. 50-277-9219 Supplier Grainger Supplier No. 19MT88
The mechanical traction extension strut is designed for precise load bearing and controlled extension. Engineered from carbon steel with a nitride finish, this robust component ensures durability and reliability in various mechanical systems. It operates as an extension-only damper with a fixed resistance design, providing stable performance across a temperature range up to 400 °F.
  • Stroke length of 10 in.
  • Constructed from carbon steel with a nitride finish
  • Fixed resistance mechanism for consistent operation
  • Maximum operating temperature of 400 °F
  • Features an extended length of 30 19/64 in. and a compressed length of 20 19/64 in.
  • Equipped with an M6 x 1 thread size