Learn More
Grainger M-STRUTS Extension Strut, Carbon Steel, 4 in. Stroke, 1 ea
Catalog No. 502779234
Encompass_Preferred
This mechanical traction extension strut is engineered from carbon steel for reliable performance in various industrial applications. Featuring a durable nitride finish, this component provides consistent force and effective damping for extension-only operations.
  • Carbon steel construction with a nitride finish
  • Designed for extension-only damping applications
  • Maximum operating temperature of 400°F
  • 4 in. stroke length
  • 15 lbf. force rating
  • M6 x 1 thread size
Catalog No. 50-277-9234 Supplier Grainger Supplier No. 19MT50
This mechanical traction extension strut is engineered from carbon steel for reliable performance in various industrial applications. Featuring a durable nitride finish, this component provides consistent force and effective damping for extension-only operations.
  • Carbon steel construction with a nitride finish
  • Designed for extension-only damping applications
  • Maximum operating temperature of 400°F
  • 4 in. stroke length
  • 15 lbf. force rating
  • M6 x 1 thread size