Learn More
Grainger Raymond Mechanical Spring Strut, Stainless Steel, 4 in. Stroke, 1 ea.
Catalog No. 502783074
Encompass_Preferred
This extension strut is a mechanical spring device designed to provide reliable, consistent, and repeatable load performance as a robust alternative to conventional gas springs. Engineered with advanced coiled spring technology and stainless steel construction, it operates effectively in harsh environments and extreme temperature conditions without the need for internal gases or seals.
  • Maintains consistent, repeatable loads with no internal gases or seals
  • Constructed from durable stainless steel
  • Designed for long-term reliability and low-friction performance
  • Operates across a wide temperature range of -40°F to 400°F
  • Features an extension-only damper type
Catalog No. 50-278-3074 Supplier Grainger Supplier No. 19MT57
This extension strut is a mechanical spring device designed to provide reliable, consistent, and repeatable load performance as a robust alternative to conventional gas springs. Engineered with advanced coiled spring technology and stainless steel construction, it operates effectively in harsh environments and extreme temperature conditions without the need for internal gases or seals.
  • Maintains consistent, repeatable loads with no internal gases or seals
  • Constructed from durable stainless steel
  • Designed for long-term reliability and low-friction performance
  • Operates across a wide temperature range of -40°F to 400°F
  • Features an extension-only damper type